Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191883.1 | NP_001178812.1 | 12 - 13 | 1144 | 59 | |
XM_006243802.2 | XP_006243864.1 | 13 - 14 | 1640 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |