Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031554.1 | NP_113742.1 | 5 - 6 | 879 | 71 | |
XM_006244072.2 | XP_006244134.1 | 4 - 5 | 981 | 71 | ||
GenBank mRNA | BC097358.1 | - | 5 - 6 | 879 | 7442221 | 71 |
L10640.1 | - | 3 - 4 | 323 | 71 | ||
M87067.1 | - | 5 - 6 | 1116 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |