Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009713.2 | NP_001009713.1 | 1 - 2 | 138 | 94 | |
GenBank mRNA | AY800277.1 | - | 1 - 2 | 137 | 94 | |
BC097482.1 | - | 1 - 2 | 138 | 7459136 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |