Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 15 - 16 | 2344 | 109 | |
XM_006243872.2 | XP_006243934.1 | 15 - 16 | 2343 | 109 | ||
XM_006243880.2 | XP_006243942.1 | 11 - 12 | 1955 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |