Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 4 - 5 | 695 | 57 | |
XM_006243872.2 | XP_006243934.1 | 4 - 5 | 694 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |