Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106851.2 | NP_001100321.1 | 2 - 3 | 353 | 146 | |
GenBank mRNA | BC167009.1 | - | 2 - 3 | 353 | 9034703 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |