Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qtrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022250.2 | NP_071586.2 | 2 - 3 | 326 | 74 | |
GenBank mRNA | AB034634.1 | - | 2 - 3 | 321 | 74 | |
BC097321.1 | - | 2 - 3 | 326 | 7387393 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |