Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057152.2 | NP_476493.2 | 4 - 5 | 1842 | 80 | |
XM_006242591.2 | XP_006242653.1 | 4 - 5 | 1888 | 80 | ||
XM_006242592.2 | XP_006242654.1 | 3 - 4 | 2389 | 80 | ||
GenBank mRNA | AF304364.1 | - | 3 - 4 | 1608 | 80 | |
FQ212383.1 | - | 4 - 5 | 1842 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |