Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139262.1 | NP_640355.1 | 1 - 2 | 72 | 70 | |
GenBank mRNA | AF187323.1 | - | 1 - 2 | 72 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |