Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037362.1 | NP_001032439.1 | 1 - 2 | 56 | 103 | |
XM_006253564.2 | XP_006253626.1 | 1 - 2 | 105 | 103 | ||
GenBank mRNA | BC087713.1 | - | 1 - 2 | 67 | 103 | |
BC107923.1 | - | 1 - 2 | 56 | 7444531 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |