Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacrgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251057.2 | XP_006251119.1 | 4 - 5 | 518 | 93 | |
XM_008770221.1 | XP_008768443.1 | 4 - 5 | 1326 | 93 | ||
XM_008770222.1 | XP_008768444.1 | 4 - 5 | 947 | 93 | ||
XM_341225.7 | XP_341226.5 | 4 - 5 | 427 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |