Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012087.1 | NP_001012087.1 | 5 - 6 | 1805 | 120 | |
XM_008761841.1 | XP_008760063.1 | 5 - 6 | 2496 | 120 | ||
XM_008761842.1 | XP_008760064.1 | 4 - 5 | 1628 | 120 | ||
XM_008761843.1 | XP_008760065.1 | 4 - 5 | 1628 | 120 | ||
XM_008761844.1 | XP_008760066.1 | 5 - 6 | 2086 | 120 | ||
GenBank mRNA | BC083842.1 | - | 5 - 6 | 1805 | 7114399 | 120 |
FQ222785.1 | - | 2 - 3 | 1375 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |