Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012086.1 | NP_001012086.1 | 2 - 3 | 269 | 120 | |
GenBank mRNA | BC088207.1 | - | 2 - 3 | 269 | 7381695 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |