Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025028.2 | NP_001020199.2 | 6 - 7 | 2135 | 76 | |
XM_008771135.1 | XP_008769357.1 | 5 - 6 | 2065 | 76 | ||
XM_008771136.1 | XP_008769358.1 | 4 - 5 | 1869 | 76 | ||
XM_008771137.1 | XP_008769359.1 | 4 - 5 | 1833 | 76 | ||
XM_008771138.1 | XP_008769360.1 | 6 - 7 | 2135 | 76 | ||
GenBank mRNA | CB613349.1 | - | 1 - 2 | 155 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |