Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034007.1 | NP_001029179.1 | 4 - 5 | 429 | 157 | |
NM_001034008.1 | NP_001029180.1 | 4 - 5 | 475 | 157 | ||
XM_006252660.2 | XP_006252722.1 | 4 - 5 | 776 | 157 | ||
GenBank mRNA | BC160817.1 | - | 4 - 5 | 454 | 7454028 | 157 |
CV119475.1 | - | 4 - 5 | 453 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |