Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034007.1 | NP_001029179.1 | 7 - 8 | 763 | 64 | |
NM_001034008.1 | NP_001029180.1 | 7 - 8 | 809 | 64 | ||
XM_006252660.2 | XP_006252722.1 | 7 - 8 | 1110 | 64 | ||
GenBank mRNA | BC160817.1 | - | 7 - 8 | 788 | 7454028 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |