Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wapal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252723.2 | XP_006252785.1 | 6 - 7 | 2218 | 106 | |
XM_006252724.2 | XP_006252786.1 | 6 - 7 | 2330 | 106 | ||
XM_006252725.2 | XP_006252787.1 | 6 - 7 | 2206 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |