Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198731.2 | NP_942026.1 | 1 - 2 | 197 | 76 | |
GenBank mRNA | BC085787.1 | - | 1 - 2 | 197 | 7108667 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |