Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168584.1 | NP_001162056.1 | 1 | 995 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |