Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257596.2 | XP_006257658.1 | 20 - 21 | 2397 | 61 | |
XM_006257597.2 | XP_006257659.1 | 20 - 21 | 3410 | 61 | ||
XM_006257599.2 | XP_006257661.1 | 20 - 21 | 3410 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Probe spans exons |