Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199095.1 | NP_954526.1 | 3 - 4 | 2580 | 102 | |
XM_006236459.1 | XP_006236521.1 | 4 - 5 | 2322 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |