Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025709.1 | NP_001020880.1 | 3 - 4 | 303 | 63 | |
GenBank mRNA | BC099162.1 | - | 3 - 4 | 303 | 7381935 | 63 |
FQ211568.1 | - | 412 | 63 | |||
FQ229403.1 | - | 3 - 4 | 288 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |