Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031725.1 | NP_113913.1 | 3 - 4 | 204 | 75 | |
XM_006240999.1 | XP_006241061.1 | 3 - 4 | 223 | 75 | ||
GenBank mRNA | AF285631.1 | - | 3 - 4 | 204 | 75 | |
BC090342.1 | - | 3 - 4 | 198 | 7304207 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |