Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108067.1 | NP_001101537.1 | 2112 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |