Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108737.1 | NP_001102207.1 | 8 - 9 | 821 | 71 | |
XM_006240977.2 | XP_006241039.1 | 8 - 9 | 1056 | 71 | ||
XM_006240978.1 | XP_006241040.1 | 8 - 9 | 843 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |