Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034112.1 | NP_001029284.1 | 2 - 3 | 245 | 113 | |
XM_006247362.2 | XP_006247424.1 | 2 - 3 | 143 | 113 | ||
GenBank mRNA | BC128702.1 | - | 2 - 3 | 279 | 7323425 | 113 |
CO560831.1 | - | 2 - 3 | 245 | 113 | ||
DQ350891.1 | - | 2 - 3 | 82 | 113 | ||
DQ350892.1 | - | 2 - 3 | 82 | 113 | ||
DQ350893.1 | - | 2 - 3 | 82 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |