Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270807.1 | NP_001257736.1 | 3 - 4 | 648 | 87 | |
NM_001270808.1 | NP_001257737.1 | 3 - 4 | 697 | 87 | ||
NM_001270809.1 | NP_001257738.1 | 3 - 4 | 598 | 87 | ||
NM_001270810.1 | NP_001257739.1 | 3 - 4 | 1284 | 87 | ||
NM_001270811.1 | NP_001257740.1 | 3 - 4 | 1634 | 87 | ||
NM_133386.3 | NP_596877.2 | 2 - 3 | 1888 | 87 | ||
XM_006247787.2 | XP_006247849.1 | 3 - 4 | 968 | 87 | ||
XM_006247788.2 | XP_006247850.1 | 3 - 4 | 479 | 87 | ||
GenBank mRNA | AB049571.1 | - | 3 - 4 | 696 | 87 | |
AB049572.1 | - | 3 - 4 | 648 | 87 | ||
AB049573.1 | - | 3 - 4 | 595 | 87 | ||
AB049574.1 | - | 3 - 4 | 1254 | 87 | ||
AB049575.1 | - | 2 - 3 | 1401 | 87 | ||
BC081738.1 | - | 2 - 3 | 1853 | 7132896 | 87 | |
DQ486894.1 | - | 3 - 4 | 436 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |