Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767917.1 | XP_008766139.1 | 4 - 5 | 488 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |