Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012570.2 | NP_036702.1 | 1 - 2 | 614 | 102 | |
GenBank mRNA | BC081841.1 | - | 1 - 2 | 498 | 7107138 | 102 |
X14044.1 | - | 1 - 2 | 504 | 102 | ||
X14223.1 | - | 1 - 2 | 477 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |