Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145782.1 | NP_665725.1 | 3 - 4 | 278 | 116 | |
GenBank mRNA | D38381.1 | - | 3 - 4 | 278 | 116 | |
X79991.1 | - | 3 - 4 | 278 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |