Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107561.1 | NP_001101031.1 | 2 - 3 | 282 | 61 | |
GenBank mRNA | X60661.1 | - | 2 - 3 | 261 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |