Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270762.1 | NP_001257691.1 | 7 - 8 | 1051 | 134 | |
NM_001270763.1 | NP_001257692.1 | 7 - 8 | 1057 | 134 | ||
NM_139105.3 | NP_620805.2 | 7 - 8 | 1066 | 134 | ||
XM_008759970.1 | XP_008758192.1 | 7 - 8 | 1615 | 134 | ||
GenBank mRNA | AB296102.1 | - | 6 - 7 | 933 | 134 | |
BC070501.1 | - | 7 - 8 | 973 | 7096144 | 134 | |
FQ233708.1 | - | 7 - 8 | 991 | 134 | ||
FQ234889.1 | - | 7 - 8 | 993 | 134 | ||
X62528.1 | - | 7 - 8 | 1051 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |