Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139093.1 | NP_620793.1 | 10 - 11 | 1321 | 63 | |
XM_006230510.2 | XP_006230572.1 | 11 - 12 | 1489 | 63 | ||
XM_006230511.2 | XP_006230573.1 | 11 - 12 | 1447 | 63 | ||
XM_006230513.2 | XP_006230575.1 | 11 - 12 | 1486 | 63 | ||
XM_006230514.2 | XP_006230576.1 | 11 - 12 | 1489 | 63 | ||
XM_006230515.2 | XP_006230577.1 | 11 - 12 | 1489 | 63 | ||
XM_006230516.2 | XP_006230578.1 | 11 - 12 | 1489 | 63 | ||
XM_006230517.2 | XP_006230579.1 | 11 - 12 | 1486 | 63 | ||
GenBank mRNA | U49057.1 | - | 10 - 11 | 1321 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |