Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081980.1 | NP_001075449.1 | 6 - 7 | 628 | 61 | |
XM_006230840.2 | XP_006230902.1 | 6 - 7 | 681 | 61 | ||
GenBank mRNA | CO564143.1 | - | 154 | 61 | ||
FQ226277.1 | - | 6 - 7 | 639 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |