Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191951.1 | NP_001178880.1 | 1 - 2 | 125 | 101 | |
XM_008760214.1 | XP_008758436.1 | 1 - 2 | 163 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |