Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135716.1 | NP_001129188.1 | 9 - 10 | 1491 | 150 | |
XM_006232515.2 | XP_006232577.1 | 9 - 10 | 1553 | 150 | ||
XM_006232516.2 | XP_006232578.1 | 8 - 9 | 1405 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |