Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106444.1 | NP_001099914.1 | 4 - 5 | 802 | 62 | |
GenBank mRNA | BC167108.1 | - | 4 - 5 | 363 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |