Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024797.1 | NP_001019968.1 | 27 - 28 | 5728 | 89 | |
XM_006232735.2 | XP_006232797.1 | 27 - 28 | 5719 | 89 | ||
XM_006232736.2 | XP_006232798.1 | 27 - 28 | 5704 | 89 | ||
XM_006232737.2 | XP_006232799.1 | 26 - 27 | 5683 | 89 | ||
XM_006232738.2 | XP_006232800.1 | 26 - 27 | 5680 | 89 | ||
XM_006232739.2 | XP_006232801.1 | 27 - 28 | 5659 | 89 | ||
XM_006232740.2 | XP_006232802.1 | 26 - 27 | 5515 | 89 | ||
XM_006232741.2 | XP_006232803.1 | 26 - 27 | 5509 | 89 | ||
XM_008761201.1 | XP_008759423.1 | 28 - 29 | 5733 | 89 | ||
XM_008761202.1 | XP_008759424.1 | 26 - 27 | 5168 | 89 | ||
GenBank mRNA | AY724781.1 | - | 27 - 28 | 5728 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |