Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762866.1 | XP_008761088.1 | 8 - 9 | 4230 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |