Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024149.1 | NP_077063.1 | 2 - 3 | 243 | 120 | |
GenBank mRNA | BC087692.1 | - | 2 - 3 | 205 | 7310424 | 120 |
L12384.1 | - | 2 - 3 | 243 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |