Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011911.1 | NP_001011911.1 | 8 - 9 | 1173 | 120 | |
XM_006251087.1 | XP_006251149.1 | 8 - 9 | 1179 | 120 | ||
GenBank mRNA | BC079008.1 | - | 8 - 9 | 1173 | 7113133 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |