Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100806.1 | NP_001094276.1 | 1 - 2 | 919 | 72 | |
GenBank mRNA | BC168678.1 | - | 1 - 2 | 919 | 5598674 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |