Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108813.2 | NP_001102283.2 | 1 - 2 | 187 | 83 | |
GenBank mRNA | BC168213.1 | - | 1 - 2 | 163 | 83 | |
FM054992.1 | - | 1 - 2 | 172 | 83 | ||
FQ214979.1 | - | 1 - 2 | 148 | 83 |
Gene Symbol: NEWGENE_1560955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773489.1 | XP_008771711.1 | 1 - 2 | 278 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |