Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sergef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229227.2 | XP_006229289.1 | 1 - 2 | 99 | 101 | |
XM_006229228.2 | XP_006229290.1 | 1 - 2 | 99 | 101 | ||
XM_006229229.2 | XP_006229291.1 | 1 - 2 | 99 | 101 | ||
XM_344891.7 | XP_344892.4 | 1 - 2 | 99 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |