Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172317.2 | NP_758528.1 | 6 - 7 | 241 | 73 | |
XM_006228820.2 | XP_006228882.1 | 6 - 7 | 481 | 73 | ||
GenBank mRNA | AW142066.1 | - | 5 - 6 | 246 | 73 | |
BG373727.1 | - | 2 - 3 | 73 | |||
CA505583.1 | - | 2 - 3 | 73 | |||
DY471861.1 | - | 6 - 7 | 613 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |