Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109544.1 | NP_001103014.1 | 3 - 4 | 366 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Recommended Assay Rattus norvegicus Probe spans exons |