Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012901.1 | NP_037033.1 | 3 - 4 | 353 | 119 | |
XM_008763759.1 | XP_008761981.1 | 3 - 4 | 439 | 119 | ||
GenBank mRNA | BC088166.1 | - | 3 - 4 | 338 | 7368246 | 119 |
S87544.1 | - | 3 - 4 | 353 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |