Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232612.2 | XP_006232674.1 | 4 - 5 | 1409 | 124 | |
GenBank mRNA | BC099090.1 | - | 4 - 5 | 694 | 124 | |
FQ216199.1 | - | 1 - 2 | 78 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |