Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106627.3 | NP_001100097.2 | 5 - 6 | 347 | 81 | |
XM_008763378.1 | XP_008761600.1 | 1 - 2 | 784 | 81 | ||
GenBank mRNA | BC160907.1 | - | 5 - 6 | 347 | 7455489 | 81 |
CV108923.1 | - | 5 - 6 | 346 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |