Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134711.1 | NP_001128183.1 | 2 - 3 | 423 | 62 | |
NM_001134712.1 | NP_001128184.1 | 2 - 3 | 259 | 62 | ||
GenBank mRNA | BC168169.1 | - | 2 - 3 | 423 | 9024666 | 62 |
EV762285.1 | - | 2 - 3 | 290 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |